Amino-PEG10-t-butyl ester 95%

BLDPBD00850797-250EA 188 EUR
BLDPBD00850797-250 BLDPBD00850797-50M BLDPBD00850797-100
Amino-PEG10-t-butyl ester 95%
Amino-PEG10-t-butyl ester

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₂₇H₅₅NO₁₂
Molekulargewicht: 585,725 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD26127812
CAS-Nummer: 1818294-42-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR