1H-Indazol-5-boronsäure 98%

BLDPBD01098688-1GEA 89 EUR
BLDPBD01098688-1G BLDPBD01098688-250 BLDPBD01098688-0.0 BLDPBD01098688-100 BLDPBD01098688-25G BLDPBD01098688-5G
1H-Indazol-5-boronsäure 98%
1H-Indazole-5-boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₇BN₂O₂
Molekulargewicht: 161,9537 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD06739151
CAS-Nummer: 338454-14-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR