3-(3,5-Dimethylisoxazol-4-yl)phenylamine 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD01183794-1GEA 404 EUR
BLDPBD01183794-1G BLDPBD01183794-250 BLDPBD01183794-100 BLDPBD01183794-50M
3-(3,5-Dimethylisoxazol-4-yl)phenylamine 98%
3-(3,5-Dimethylisoxazol-4-yl)phenylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₁H₁₂N₂O
Molekulargewicht: 188,226 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD20701923
CAS-Nummer: 875628-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR