N-Benzylidenanilin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD01290376-5GEA 27 EUR
BLDPBD01290376-5G BLDPBD01290376-25G BLDPBD01290376-100
N-Benzylidenanilin 98%
N-Benzylidenanilin

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₁₁N
Molekulargewicht: 181,24 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003027
CAS-Nummer: 538-51-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR