(R)-(-)-3-Amino-1-Boc-piperidin 97%
|
|
BLDPBD0329-25GEA
72
EUR
BLDPBD0329-25G
BLDPBD0329-100G
BLDPBD0329-5G
BLDPBD0329-10G
BLDPBD0329-1G
BLDPBD0329-500G
(R)-(-)-3-Amino-1-Boc-piperidin 97%
(R)-(-)-3-Amino-1-Boc-piperidin
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₁₀H₂₀N₂O₂ Molekulargewicht: 200,28 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03094717 CAS-Nummer: 188111-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...