(±)-3-(Boc-amino)piperidin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD0338-100GEA 336 EUR
BLDPBD0338-100G BLDPBD0338-1G BLDPBD0338-25G BLDPBD0338-5G BLDPBD0338-10G BLDPBD0338-500G
(±)-3-(Boc-amino)piperidin 98%
(±)-3-(Boc-amino)piperidin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₀H₂₀N₂O₂
Molekulargewicht: 200,28 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03839941
CAS-Nummer: 172603-05-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR