4,5-Dimethoxyisatosäureanhydrid 97%
|
|
BLDPBD0475-250MGEA
17
EUR
BLDPBD0475-250MG
BLDPBD0475-5G
BLDPBD0475-10G
BLDPBD0475-1G
BLDPBD0475-25G
4,5-Dimethoxyisatosäureanhydrid 97%
4,5-Dimethoxyisatosäureanhydrid
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₁₀H₉NO₅ Molekulargewicht: 223,1800 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03426390 CAS-Nummer: 20197-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...