4-tert-Butylphenylisothiocyanat 98%
|
|
BLDPBD0576-25GEA
165
EUR
BLDPBD0576-25G
BLDPBD0576-5G
BLDPBD0576-1G
4-tert-Butylphenylisothiocyanat 98%
4-tert-Butylphenylisothiocyanat
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₁₁H₁₃NS Molekulargewicht: 191,2926 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00022059 CAS-Nummer: 19241-24-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...