4-Isobutylbenzoesäure 95%

BLDPBD0599-1GEA 56 EUR
BLDPBD0599-1G BLDPBD0599-250MG BLDPBD0599-5G
4-Isobutylbenzoesäure 95%
4-Isobutylbenzoesäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₄O₂
Molekulargewicht: 178,2277 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00191658
CAS-Nummer: 38861-88-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR