4-(Thiophen-2-yl)benzoic acid 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD0705-1GEA 23 EUR
BLDPBD0705-1G BLDPBD0705-5G BLDPBD0705-250MG
4-(Thiophen-2-yl)benzoic acid 98%
4-(Thiophen-2-yl)benzoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₁H₈O₂S
Molekulargewicht: 204,25 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00690742
CAS-Nummer: 29886-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR