5-(3-Bromphenyl)oxazol 98%

BLDPBD100798-1GEA 17 EUR
BLDPBD100798-1G BLDPBD100798-25G BLDPBD100798-5G
5-(3-Bromphenyl)oxazol 98%
5-(3-Bromophenyl)oxazole

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₆BrNO
Molekulargewicht: 224,06 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD05668970
CAS-Nummer: 243455-57-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR