5-(4-Bromphenyl)oxazol 98%

BLDPBD100910-1GEA 18 EUR
BLDPBD100910-1G BLDPBD100910-25G BLDPBD100910-5G
5-(4-Bromphenyl)oxazol 98%
5-(4-Bromophenyl)oxazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₆BrNO
Molekulargewicht: 224,0540 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02682015
CAS-Nummer: 72571-06-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR