N-Boc-ethylendiamin 97%

Lieferant: BLD PHARMATECH GMBH

Synonyme: t-boc-N-EDA

BLDPBD10107-25GEA 15 EUR
BLDPBD10107-25G BLDPBD10107-10G BLDPBD10107-5G
N-Boc-ethylendiamin 97%
N-Boc-ethylendiamin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₁₆N₂O₂
Molekulargewicht: 160,22 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00191871
CAS-Nummer: 57260-73-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR