5,6-Dichlor-2-methylbenzimidazol 98%

BLDPBD101655-250MGEA 13 EUR
BLDPBD101655-250MG BLDPBD101655-25G BLDPBD101655-1G BLDPBD101655-5G
5,6-Dichlor-2-methylbenzimidazol 98%
5,6-Dichloro-2-methylbenzimidazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆Cl₂N₂
Molekulargewicht: 201,0500 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00276973
CAS-Nummer: 6478-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR