5-Amino-1-methyl-2-oxoindoline 98%
|
|
BLDPBD102087-100MGEA
74
EUR
BLDPBD102087-100MG
BLDPBD102087-5G
BLDPBD102087-1G
BLDPBD102087-250MG
5-Amino-1-methyl-2-oxoindoline 98%
5-Amino-1-methyl-2-oxoindoline
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₉H₁₀N₂O Molekulargewicht: 162,189 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09702413 CAS-Nummer: 20870-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...