5-Amino-2-methylbenzolsulfonamid 95%

BLDPBD102190-100GEA 68 EUR
BLDPBD102190-100G BLDPBD102190-5G BLDPBD102190-1G BLDPBD102190-25G BLDPBD102190-10G BLDPBD102190-0.025
5-Amino-2-methylbenzolsulfonamid 95%
5-Amino-2-methylbenzolsulfonamid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₁₀N₂O₂S
Molekulargewicht: 186,23 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06681071
CAS-Nummer: 6973-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR