Ethyl-5-amino-3-methyl-1H-pyrazol-4-carboxylat 98%

BLDPBD102249-1GEA 77 EUR
BLDPBD102249-1G BLDPBD102249-5G BLDPBD102249-250MG
Ethyl-5-amino-3-methyl-1H-pyrazol-4-carboxylat 98%
Ethyl-5-amino-3-methyl-1H-pyrazol-4-carboxylat

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₁₁N₃O₂
Molekulargewicht: 169,18 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01210846
CAS-Nummer: 23286-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR