5-Brom-1-chlorisochinolin 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD102537-1GEA 74 EUR
BLDPBD102537-1G BLDPBD102537-5G BLDPBD102537-25G BLDPBD102537-250MG BLDPBD102537-10G BLDPBD102537-100G BLDPBD102537-100MG
5-Brom-1-chlorisochinolin 95%
5-Bromo-1-chloroisoquinoline

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₅BrClN
Molekulargewicht: 242,4997 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08703278
CAS-Nummer: 34551-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR