2-Chlor-3-fluorbenzaldehyd 95%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD10267-5GEA
25
EUR
BLDPBD10267-5G
BLDPBD10267-1G
BLDPBD10267-100G
BLDPBD10267-10G
BLDPBD10267-25G
2-Chlor-3-fluorbenzaldehyd 95%
2-Chloro-3-fluorobenzaldehyde
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₇H₄ClFO Molekulargewicht: 158,56 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD07782053 CAS-Nummer: 96516-31-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...