5-Brom-2-fluor-4-methylpyridin 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD102704-1GEA 6 EUR
BLDPBD102704-1G BLDPBD102704-25G BLDPBD102704-500G BLDPBD102704-100G BLDPBD102704-5G BLDPBD102704-10G
5-Brom-2-fluor-4-methylpyridin 97%
5-Brom-2-fluor-4-methylpyridin

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₆H₅BrFN
Molekulargewicht: 190,01 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09839213
CAS-Nummer: 864830-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR