5-Brom-2-(methylthio)pyridin 98%

BLDPBD102820-250MGEA 17 EUR
BLDPBD102820-250MG BLDPBD102820-5G BLDPBD102820-1G BLDPBD102820-25G
5-Brom-2-(methylthio)pyridin 98%
5-Brom-2-(methylthio)pyridin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₆BrNS
Molekulargewicht: 204,0875 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03411571
CAS-Nummer: 51933-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR