6-Chloroisatoic anhydride 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD103494-5GEA 188 EUR
BLDPBD103494-5G BLDPBD103494-250MG BLDPBD103494-1G BLDPBD103494-100MG
6-Chloroisatoic anhydride 98%
6-Chloroisatoic anhydride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₄ClNO₃
Molekulargewicht: 197,575 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09032504
CAS-Nummer: 20829-96-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR