6-Brompicolinamid 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD108576-10GEA 230 EUR
BLDPBD108576-10G BLDPBD108576-1G BLDPBD108576-25G BLDPBD108576-250MG BLDPBD108576-5G
6-Brompicolinamid 98%
6-Brompicolinamid

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₅BrN₂O
Molekulargewicht: 201,02 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00234154
CAS-Nummer: 25194-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR