2-Brom-2',4'-difluoracetophenon 98%

BLDPBD10859-25GEA 74 EUR
BLDPBD10859-25G BLDPBD10859-5G BLDPBD10859-0.025G
2-Brom-2',4'-difluoracetophenon 98%
2-Brom-2',4'-difluoracetophenon

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₈H₅BrF₂O
Molekulargewicht: 235,03 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00142822
CAS-Nummer: 102429-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR