6-Chloropyridine-2-carboxamide 98%

BLDPBD109008-250MGEA 57 EUR
BLDPBD109008-250MG BLDPBD109008-1G BLDPBD109008-5G BLDPBD109008-100MG
6-Chloropyridine-2-carboxamide 98%
6-Chloropyridine-2-carboxamide

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₅ClN₂O
Molekulargewicht: 156,569 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00708892
CAS-Nummer: 70593-61-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR