6-Chlorchinoxalin 98%

BLDPBD109023-5GEA 29 EUR
BLDPBD109023-5G BLDPBD109023-1G
6-Chlorchinoxalin 98%
6-Chloroquinoxaline

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₅ClN₂
Molekulargewicht: 164,5917 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00047594
CAS-Nummer: 5448-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR