3',5'-Difluor-2'-hydroxyacetophenon 98%

BLDPBD10959-10GEA 128 EUR
BLDPBD10959-10G BLDPBD10959-250MG BLDPBD10959-1G BLDPBD10959-25G BLDPBD10959-5G
3',5'-Difluor-2'-hydroxyacetophenon 98%
3',5'-Difluor-2'-hydroxyacetophenon

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆F₂O₂
Molekulargewicht: 172,13 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00042476
CAS-Nummer: 140675-42-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR