6-Morpholino-3-pyridinamin 97%

BLDPBD109797-5GEA 260 EUR
BLDPBD109797-5G BLDPBD109797-250MG BLDPBD109797-1G
6-Morpholino-3-pyridinamin 97%
6-Morpholino-3-pyridinamin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₁₃N₃O
Molekulargewicht: 179,22 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00119562
CAS-Nummer: 52023-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR