1H-Indazol-3-carbonsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD10995-10GEA 9 EUR
BLDPBD10995-10G BLDPBD10995-500G BLDPBD10995-100G BLDPBD10995-5G BLDPBD10995-25G
1H-Indazol-3-carbonsäure 98%
1H-Indazole-3-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆N₂O₂
Molekulargewicht: 162,15 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00211066
CAS-Nummer: 4498-67-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR