7-Fluor-1H-indazol 98%

BLDPBD110578-25GEA 189 EUR
BLDPBD110578-25G BLDPBD110578-100G BLDPBD110578-5G BLDPBD110578-1G BLDPBD110578-10G
7-Fluor-1H-indazol 98%
7-Fluoro-1H-indazole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₅FN₂
Molekulargewicht: 136,1264 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09026976
CAS-Nummer: 341-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR