4-(2-Bromoethyl)phenol 98%

BLDPBD11158-25GEA 207 EUR
BLDPBD11158-25G BLDPBD11158-1G BLDPBD11158-250MG BLDPBD11158-100G BLDPBD11158-10G BLDPBD11158-5G
4-(2-Bromoethyl)phenol 98%
4-(2-Bromoethyl)phenol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₈H₉BrO
Molekulargewicht: 201,06 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00129679
CAS-Nummer: 14140-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR