Methyl-3-amino-4-methylbenzoat 97%

BLDPBD11292-1000GEA 311 EUR
BLDPBD11292-1000G BLDPBD11292-10G BLDPBD11292-25G BLDPBD11292-500G BLDPBD11292-100G
Methyl-3-amino-4-methylbenzoat 97%
Methyl-3-amino-4-methylbenzoat

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₁₁NO₂
Molekulargewicht: 165,19 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00025206
CAS-Nummer: 18595-18-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR