2,7-Chinolindiol 95%

BLDPBD11588-5GEA 53 EUR
BLDPBD11588-5G BLDPBD11588-10G BLDPBD11588-1G BLDPBD11588-25G
2,7-Chinolindiol 95%
2,7-Chinolindiol

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₇NO₂
Molekulargewicht: 161,157 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04117979
CAS-Nummer: 70500-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR