Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')nickel(II) 98%
|
|
BLDPBD116834-250MGEA
38
EUR
BLDPBD116834-250MG
BLDPBD116834-1G
BLDPBD116834-5G
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')nickel(II) 98%
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')nickel(II)
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₂₂H₃₈NiO₄ Molekulargewicht: 425,23 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00192348 CAS-Nummer: 14481-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...