9,10-Bis(2-naphthyl)anthracene-2-ylboronic acid 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD118638-25GEA 529 EUR
BLDPBD118638-25G BLDPBD118638-5G BLDPBD118638-1G BLDPBD118638-250MG
9,10-Bis(2-naphthyl)anthracene-2-ylboronic acid 95%
9,10-Bis(2-naphthyl)anthracene-2-ylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₃₄H₂₃BO₂
Molekulargewicht: 474,356 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11045063
CAS-Nummer: 867044-28-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR