3'-Acetamidoacetophenon 98%
|
|
BLDPBD11929-100GEA
90
EUR
BLDPBD11929-100G
BLDPBD11929-5G
BLDPBD11929-500G
BLDPBD11929-25G
3'-Acetamidoacetophenon 98%
3'-Acetamidoacetophenon
Storage condition: Sealed in dry, Room Temperature
Formel:
C₁₀H₁₁NO₂ Molekulargewicht: 177,20 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00032278 CAS-Nummer: 7463-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...