Ethylisodehydroacetat 99%

BLDPBD12067-5GEA 113 EUR
BLDPBD12067-5G BLDPBD12067-250MG BLDPBD12067-25G BLDPBD12067-1G
Ethylisodehydroacetat 99%
Ethylisodehydroacetat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₂O₄
Molekulargewicht: 196,1999 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00006643
CAS-Nummer: 3385-34-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR