2-Methoxybenzeneboronic acid, propanediol cyclic ester 98%

BLDPBD12085-250MGEA 11 EUR
BLDPBD12085-250MG BLDPBD12085-5G BLDPBD12085-1G
2-Methoxybenzeneboronic acid, propanediol cyclic ester 98%
2-Methoxybenzeneboronic acid, propanediol cyclic ester

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₃BO₃
Molekulargewicht: 192,019 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06659934
CAS-Nummer: 141522-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR