(Chlormethyl)dimethylphenylsilan 97%
|
|
BLDPBD120884-1GEA
11
EUR
BLDPBD120884-1G
BLDPBD120884-5G
BLDPBD120884-0.025
BLDPBD120884-25G
BLDPBD120884-250MG
(Chlormethyl)dimethylphenylsilan 97%
(Chlormethyl)dimethylphenylsilan
Storage condition: Sealed in dry, Room Temperature
Formel:
C₉H₁₃ClSi Molekulargewicht: 184,7400 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00040858 CAS-Nummer: 1833-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...