Zimtsäurechlorid 97% (predominantly trans isomers)

BLDPBD121362-25GEA 6 EUR
BLDPBD121362-25G
Zimtsäurechlorid 97% (predominantly trans isomers)
Zimtsäurechlorid

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₇ClO
Molekulargewicht: 166,6043 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00000732
CAS-Nummer: 102-92-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR