Cytidin-5'-triphosphorsäure Dinatriumsalz (CTP-Na2) 97%

BLDPBD123181-25GEA 339 EUR
BLDPBD123181-25G BLDPBD123181-5G BLDPBD123181-250MG BLDPBD123181-1G
Cytidin-5'-triphosphorsäure Dinatriumsalz (CTP-Na2) 97%
Cytidin-5'-triphosphorsäure Dinatriumsalz (CTP-Na2)

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₉H₁₄N₃Na₂O₁₄P₃
Molekulargewicht: 527,12 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD00078193
CAS-Nummer: 36051-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR