D(+)-Cellobiose 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD123228-5GEA 9 EUR
BLDPBD123228-5G BLDPBD123228-25G BLDPBD123228-0.025 BLDPBD123228-100G
D(+)-Cellobiose 98%
D(+)-Cellobiose

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₂H₂₂O₁₁
Molekulargewicht: 342,30 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00136034
CAS-Nummer: 528-50-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR