D(+)-Turanose 98%

BLDPBD123258-1GEA 59 EUR
BLDPBD123258-1G BLDPBD123258-250MG
D(+)-Turanose 98%
D(+)-Turanose

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₂₂O₁₁
Molekulargewicht: 342,30 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00006606
CAS-Nummer: 547-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR