Isochinolin-1-carbonitril 95%

BLDPBD12536-25GEA 498 EUR
BLDPBD12536-25G BLDPBD12536-100G BLDPBD12536-5G BLDPBD12536-1G BLDPBD12536-250MG
Isochinolin-1-carbonitril 95%
Isochinolin-1-carbonitril

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₆N₂
Molekulargewicht: 154,17 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00134166
CAS-Nummer: 1198-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR