3-Chlor-6-phenylpyridazin 96%
|
|
BLDPBD12577-10GEA
256
EUR
BLDPBD12577-10G
BLDPBD12577-25G
BLDPBD12577-5G
BLDPBD12577-250MG
BLDPBD12577-1G
3-Chlor-6-phenylpyridazin 96%
3-Chloro-6-phenylpyridazine
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₁₀H₇ClN₂ Molekulargewicht: 190,6300 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00075253 CAS-Nummer: 20375-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...