Diphenylacetaldehyd 95%

BLDPBD125854-25GEA 251 EUR
BLDPBD125854-25G BLDPBD125854-10G BLDPBD125854-5G BLDPBD125854-1G
Diphenylacetaldehyd 95%
Diphenylacetaldehyd

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₄H₁₂O
Molekulargewicht: 196,2400 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00006972
CAS-Nummer: 947-91-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR