Isochinolin-5-carbaldehyd 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD12615-100MGEA 9 EUR
BLDPBD12615-100MG BLDPBD12615-5G BLDPBD12615-250MG BLDPBD12615-1G
Isochinolin-5-carbaldehyd 98%
Isochinolin-5-carbaldehyd

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₀H₇NO
Molekulargewicht: 157,17 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03412483
CAS-Nummer: 80278-67-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR