Isochinolin-1-ylmethanol 95%

BLDPBD12845-250MGEA 111 EUR
BLDPBD12845-250MG BLDPBD12845-100MG
Isochinolin-1-ylmethanol 95%
Isochinolin-1-ylmethanol
Formel: C₁₀H₉NO
Molekulargewicht: 159,1846 g/mol
MDL Number: MFCD03086097
CAS-Nummer: 27311-63-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR