4-(Ethoxycarbonyl)phenylboronsäurepinakolester 95%

BLDPBD128732-25GEA 370 EUR
BLDPBD128732-25G BLDPBD128732-1G BLDPBD128732-10G BLDPBD128732-5G
4-(Ethoxycarbonyl)phenylboronsäurepinakolester 95%
4-(Ethoxycarbonyl)phenylboronsäurepinakolester

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₅H₂₁BO₄
Molekulargewicht: 276,136 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD03453659
CAS-Nummer: 195062-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR