Ethyl-5-amino-1-(3-chlorphenyl)-1H-pyrazol-4-carboxylat 95+%

BLDPBD129029-1GEA 97 EUR
BLDPBD129029-1G BLDPBD129029-250MG BLDPBD129029-5G
Ethyl-5-amino-1-(3-chlorphenyl)-1H-pyrazol-4-carboxylat 95+%
Ethyl-5-amino-1-(3-chlorphenyl)-1H-pyrazol-4-carboxylat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₂ClN₃O₂
Molekulargewicht: 265,7000 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00973940
CAS-Nummer: 15001-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR